ENA497680375 Enamine

[1-(2-bromoethoxy)propyl]benzene

  • MDL number MFCD28132779
  • Cas Number : 1824054-90-1
  • Molecular Weight : 243.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263577
Availability details for
Quantity
  • Availability
  • Add to Cart