ENA497680376 Enamine

tert-butyl 3-hydroxy-3-(naphthalen-1-yl)azetidine-1-carboxylate

  • MDL number MFCD29907089
  • Cas Number : 1696848-61-9
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264133
Availability details for
Quantity
  • Availability
  • Add to Cart