ENA497680378 Enamine

1-(2,4-difluorophenyl)-5-methyl-1H-pyrazol-4-amine

  • MDL number MFCD16042716
  • Cas Number : 1245007-35-5
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264156
Availability details for
Quantity
  • Availability
  • Add to Cart