ENA497680379 Enamine

2-(2-bromophenyl)-2-methoxyacetonitrile

  • MDL number MFCD21684370
  • Cas Number : 1415388-63-4
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264164
Availability details for
Quantity
  • Availability
  • Add to Cart