ENA497680390 Enamine

4H,5H,6H,7H-[1,2]oxazolo[4,3-c]pyridine hydrochloride

  • MDL number MFCD28895348
  • Cas Number : 1955493-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-264247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264247
Availability details for
Quantity
  • Availability
  • Add to Cart