ENA497680397 Enamine

3-(3-fluorophenyl)-3-methylpyrrolidine

  • MDL number MFCD16737832
  • Cas Number : 1247573-87-0
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263462

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-263462
pictograms GHS05,GHS07
hstatements H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart