ENA497680412 Enamine

2-chloro-4-(3-fluorophenyl)-1,3,5-triazine

  • MDL number MFCD11052418
  • Cas Number : 1053658-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-264292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264292
Availability details for
Quantity
  • Availability
  • Add to Cart