ENA497680415 Enamine

3,4-dihydro-1H-spiro[naphthalene-2,4'-piperidine]-1-one hydrochloride

  • MDL number MFCD18711482
  • Cas Number : 312600-59-2
  • Molecular Weight : 251.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263589
Availability details for
Quantity
  • Availability
  • Add to Cart