ENA497680416 Enamine

dicyclohexylmethanol

  • MDL number MFCD00001474
  • Cas Number : 4453-82-1
  • Molecular Weight : 196.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-64
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264331
Availability details for
Quantity
  • Availability
  • Add to Cart