ENA497680428 Enamine

2-chloroethyl N-(2-bromophenyl)carbamate

  • MDL number MFCD01006616
  • Cas Number : 99548-44-4
  • Molecular Weight : 278.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264496
Availability details for
Quantity
  • Availability
  • Add to Cart