ENA497680433 Enamine

2-amino-3,4,5,6-tetrahydropyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD20645214
  • Cas Number : 146422-39-1
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264525
Availability details for
Quantity
  • Availability
  • Add to Cart