ENA497680454 Enamine

3-[(2-fluorophenyl)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD11856679
  • Cas Number : 1159542-76-3
  • Molecular Weight : 232.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263651
Availability details for
Quantity
  • Availability
  • Add to Cart