ENA497680468 Enamine

5-oxo-4-azatricyclo[4.2.1.0,3,7]nonane-3-carboxylic acid

  • MDL number MFCD24515724
  • Cas Number : 286456-64-2
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264713
Availability details for
Quantity
  • Availability
  • Add to Cart