ENA497680474 Enamine

(1R)-1-(1-methyl-1H-1,2,3-triazol-5-yl)ethan-1-ol

  • MDL number MFCD29907111
  • Cas Number : 1867669-43-9
  • Molecular Weight : 127.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264729
Availability details for
Quantity
  • Availability
  • Add to Cart