ENA497680479 Enamine

2-{1-[(prop-2-en-1-ylsulfanyl)methyl]cyclopropyl}acetic acid

  • MDL number MFCD17012827
  • Cas Number : 1001907-54-5
  • Molecular Weight : 186.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263660
Availability details for
Quantity
  • Availability
  • Add to Cart