ENA497681018 Enamine

5-[(tert-butoxy)carbonyl]-6-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2-carboxylic acid

  • MDL number MFCD29046120
  • Cas Number : 1955548-24-9
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265301
Availability details for
Quantity
  • Availability
  • Add to Cart