ENA497681105 Enamine

6-methyl-4H,5H-[1,2,3]triazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD29045814
  • Cas Number : 1936285-98-1
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265388
Availability details for
Quantity
  • Availability
  • Add to Cart