ENA497681132 Enamine

2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridin-3-amine

  • MDL number MFCD00278361
  • Cas Number : 60639-28-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265415
Availability details for
Quantity
  • Availability
  • Add to Cart