ENA497681406 Enamine

2-(azetidin-3-yloxy)acetic acid; trifluoroacetic acid

  • MDL number MFCD29907119
  • Cas Number : 1909327-16-7
  • Molecular Weight : 245.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265953
Availability details for
Quantity
  • Availability
  • Add to Cart