ENA497681407 Enamine

1-(2,4-difluorophenyl)-5-methyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD29907120
  • Cas Number : 1909335-81-4
  • Molecular Weight : 245.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265955
Availability details for
Quantity
  • Availability
  • Add to Cart