ENA497681413 Enamine

2-({4H,5H,6H,7H-pyrazolo[1,5-a]pyridin-2-yl}methyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD29907126
  • Cas Number : 1909320-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266007
Availability details for
Quantity
  • Availability
  • Add to Cart