ENA497681414 Enamine

2-amino-N-ethoxy-N-methylacetamide hydrochloride

  • MDL number MFCD29907128
  • Cas Number : 1909313-73-0
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266024
Availability details for
Quantity
  • Availability
  • Add to Cart