ENA497681436 Enamine

N-{5-azaspiro[3.5]nonan-8-yl}butanamide hydrochloride

  • MDL number MFCD29907154
  • Cas Number : 1909337-81-0
  • Molecular Weight : 246.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266242
Availability details for
Quantity
  • Availability
  • Add to Cart