ENA497681443 Enamine

5-chloro-6-[(3-chlorophenyl)methoxy]pyridine-3-carbonitrile

  • MDL number MFCD29907161
  • Cas Number : 1909327-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266321
Availability details for
Quantity
  • Availability
  • Add to Cart