ENA497681456 Enamine

3-(2,6-difluorophenyl)-1-methyl-1H-1,2,4-triazol-5-amine hydrobromide

  • MDL number MFCD29907180
  • Cas Number : 1909327-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266462
Availability details for
Quantity
  • Availability
  • Add to Cart