ENA497681491 Enamine

1,1-difluoro-5-azaspiro[2.3]hexane; trifluoroacetic acid

  • MDL number MFCD29907194
  • Cas Number : 1909327-96-3
  • Molecular Weight : 233.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266626
Availability details for
Quantity
  • Availability
  • Add to Cart