ENA497681492 Enamine

2-(chloromethyl)azepane hydrochloride

  • MDL number MFCD29907196
  • Cas Number : 19168-73-1
  • Molecular Weight : 184.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266631
Availability details for
Quantity
  • Availability
  • Add to Cart