ENA497681503 Enamine

(2-bromo-6-chlorophenyl)hydrazine dihydrochloride

  • MDL number MFCD29907208
  • Cas Number : 1909312-32-8
  • Molecular Weight : 294.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266874
Availability details for
Quantity
  • Availability
  • Add to Cart