ENA497681508 Enamine

4-(3,5-dimethyl-1H-pyrazol-1-yl)benzene-1-carboximidamide; acetic acid

  • MDL number MFCD29907212
  • Cas Number : 1909314-25-5
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CH3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266895
Availability details for
Quantity
  • Availability
  • Add to Cart