ENA497681516 Enamine

3-(aminomethyl)-1,1-difluoropentane hydrochloride

  • MDL number MFCD29907221
  • Cas Number : 1909305-91-4
  • Molecular Weight : 173.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267028
Availability details for
Quantity
  • Availability
  • Add to Cart