ENA497681538 Enamine

2-methyl-3-(pyridin-2-yl)prop-2-enoic acid hydrochloride

  • MDL number MFCD29907257
  • Cas Number : 1909358-81-1
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267231
Availability details for
Quantity
  • Availability
  • Add to Cart