ENA497681548 Enamine

propan-2-yl 3-amino-4-methylpentanoate hydrochloride

  • MDL number MFCD29907270
  • Cas Number : 1909306-11-1
  • Molecular Weight : 209.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267333
Availability details for
Quantity
  • Availability
  • Add to Cart