ENA497681551 Enamine

N-(2,3-dihydro-1H-isoindol-5-yl)-2,2,2-trifluoroacetamide hydrochloride

  • MDL number MFCD29907273
  • Cas Number : 1909328-14-8
  • Molecular Weight : 266.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267344
Availability details for
Quantity
  • Availability
  • Add to Cart