ENA497681552 Enamine

N-methylthiophen-3-amine hydrochloride

  • MDL number MFCD28978315
  • Cas Number : 1909337-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-267346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267346
Availability details for
Quantity
  • Availability
  • Add to Cart