ENA497681556 Enamine

7-bromo-2,3,3-trimethyl-3H-indole

  • MDL number MFCD29907280
  • Cas Number : 1701461-00-8
  • Molecular Weight : 238.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267380
Availability details for
Quantity
  • Availability
  • Add to Cart