ENA497681751 Enamine

ethyl 3-[(propan-2-yl)amino]propanoate

  • MDL number MFCD01475850
  • Cas Number : 16217-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-265865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265865
Availability details for
Quantity
  • Availability
  • Add to Cart