ENA497681795 Enamine

ethyl 2-methyl-3-(pyridin-3-yl)prop-2-enoate

  • MDL number MFCD26895431
  • Cas Number : 172656-39-2
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-265978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-265978
Availability details for
Quantity
  • Availability
  • Add to Cart