ENA497681802 Enamine

2,6-dichloro-3-(chloromethyl)quinoline

  • MDL number MFCD09787656
  • Cas Number : 948290-83-3
  • Molecular Weight : 246.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266008
Availability details for
Quantity
  • Availability
  • Add to Cart