ENA497681813 Enamine

ethyl 7-oxo-6H,7H-[1,2]thiazolo[4,3-d]pyrimidine-3-carboxylate

  • MDL number MFCD02598861
  • Cas Number : 54968-79-5
  • Molecular Weight : 225.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266035
Availability details for
Quantity
  • Availability
  • Add to Cart