ENA497681814 Enamine

methyl 2-amino-6-bromopyridine-3-carboxylate

  • MDL number MFCD19105143
  • Cas Number : 1227048-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266037
Availability details for
Quantity
  • Availability
  • Add to Cart