ENA497681818 Enamine

methyl 3-(benzenesulfonyl)prop-2-enoate

  • Cas Number : 63068-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266044
Availability details for
Quantity
  • Availability
  • Add to Cart