ENA497681839 Enamine

N-methyl-2-[2-(methylamino)phenyl]aniline

  • MDL number MFCD24550561
  • Cas Number : 173044-26-3
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266080
Availability details for
Quantity
  • Availability
  • Add to Cart