ENA497681842 Enamine

4-(4-ethylphenyl)-1H-imidazol-2-amine

  • MDL number MFCD11901319
  • Cas Number : 1176053-13-6
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266085
Availability details for
Quantity
  • Availability
  • Add to Cart