ENA497681848 Enamine

2-ethyl-2-methylcyclopentan-1-one

  • MDL number MFCD24692946
  • Cas Number : 88726-48-1
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266094
Availability details for
Quantity
  • Availability
  • Add to Cart