ENA497681862 Enamine

tert-butyl 2-(methylamino)butanoate

  • MDL number MFCD16113703
  • Cas Number : 1248382-70-8
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266116
Availability details for
Quantity
  • Availability
  • Add to Cart