ENA497681879 Enamine

1-(1-methyl-1H-pyrazol-3-yl)piperidin-4-one

  • MDL number MFCD23766992
  • Cas Number : 1538024-17-7
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266192
Availability details for
Quantity
  • Availability
  • Add to Cart