ENA497681886 Enamine

methyl 4-(2-phenylethynyl)benzoate

  • MDL number MFCD06203959
  • Cas Number : 42497-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266211
Availability details for
Quantity
  • Availability
  • Add to Cart