ENA497681888 Enamine

2-(1-methoxyethyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD26821575
  • Cas Number : 1566576-18-8
  • Molecular Weight : 171.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266218
Availability details for
Quantity
  • Availability
  • Add to Cart