ENA497681890 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-4-methanesulfonylbutanoic acid

  • MDL number MFCD09811301
  • Cas Number : 1016677-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266227
Availability details for
Quantity
  • Availability
  • Add to Cart