ENA497681897 Enamine

1-[2-(1H-indol-3-yl)ethyl]-3-(prop-2-en-1-yl)urea

  • MDL number MFCD11784039
  • Cas Number : 1110820-96-6
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266244

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P272,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P333+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-266244
pictograms GHS07
hstatements H302,H315,H317,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart